Skip to Content
Merck
All Photos(1)

Key Documents

46984

Supelco

Trimethoprim

VETRANAL®, analytical standard

Synonym(s):

2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
Beilstein:
625127
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

application(s)

clinical testing

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC

InChI

1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)

InChI key

IEDVJHCEMCRBQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: pyrimidine

Application

Primarily used as an antibacterial agent. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Grape et al.
Antimicrobial agents and chemotherapy, 51(5), 1863-1864 (2007-02-20)
Two Escherichia coli isolates resistant to trimethoprim but negative for integrons carried two new resistance genes, dfrA24 and dfrA26, remotely similar to one another and to the cassette-independent genes dfrA8 and dfrA9. The dfrA24 gene was not associated with known
Kyuho Han et al.
Nature methods, 11(1), 86-93 (2013-11-12)
Protein concentrations are often regulated by dynamic changes in translation rates. Nevertheless, it has been challenging to directly monitor changes in translation in living cells. We have developed a reporter system to measure real-time changes of translation rates in human
Christine Manyando et al.
PloS one, 9(5), e96017-e96017 (2014-05-17)
Antibiotic therapy during pregnancy may be beneficial and impacts positively on the reduction of adverse pregnancy outcomes. No studies have been done so far on the effects of daily Co-trimoxazole (CTX) prophylaxis on birth outcomes. A phase 3b randomized trial
O Sköld
Veterinary research, 32(3-4), 261-273 (2001-07-04)
Sulfonamides and trimethoprim have been used for many decades as efficient and inexpensive antibacterial agents for animals and man. Resistance to both has, however, spread extensively and rapidly. This is mainly due to the horizontal spread of resistance genes, expressing
Sohei Harada et al.
Antimicrobial agents and chemotherapy, 54(9), 3545-3550 (2010-06-23)
Integrating conjugative elements (ICEs) are mobile genetic elements that can transfer from the chromosome of a host to the chromosome of a new host through the process of excision, conjugation, and integration. Although SXT/R391-related ICEs, originally demonstrated in Vibrio cholerae

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service