Skip to Content
Merck
All Photos(1)

Key Documents

46412

Supelco

Medroxyprogesterone 17-acetate

VETRANAL®, analytical standard

Synonym(s):

17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
Beilstein:
2066112
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

206-207 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(C)=O)C(C)=O

InChI

1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1

InChI key

PSGAAPLEWMOORI-PEINSRQWSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Jeong-Yeol Park et al.
Obstetrics and gynecology, 121(1), 136-142 (2012-12-25)
To analyze pregnancy outcomes in young women with stage IA, grade 1 endometrioid adenocarcinoma of the uterus after successful fertility-sparing management using progestin. We reviewed the medical records of 141 women with stage IA, grade 1 endometrioid adenocarcinoma of the
JoAnn E Manson et al.
JAMA, 310(13), 1353-1368 (2013-10-03)
Menopausal hormone therapy continues in clinical use but questions remain regarding its risks and benefits for chronic disease prevention. To report a comprehensive, integrated overview of findings from the 2 Women's Health Initiative (WHI) hormone therapy trials with extended postintervention
Richard P H Huijbregts et al.
Endocrinology, 154(3), 1282-1295 (2013-01-29)
Recent observational studies indicate an association between the use of hormonal contraceptives and acquisition and transmission of HIV-1. The biological and immunological mechanisms underlying the observed association are unknown. Depot medroxyprogesterone acetate (DMPA) is a progestin-only injectable contraceptive that is
Robert A Wild et al.
Menopause (New York, N.Y.), 20(3), 254-260 (2013-02-26)
Our objective was to determine whether metabolic syndrome (MetS) or its components modified the effect of hormone therapy (HT) on the risk of coronary heart disease (CHD) events in the Women's Health Initiative clinical trials. We performed a nested case-control
Léanie Kleynhans et al.
Infection and immunity, 81(4), 1234-1244 (2013-02-06)
The contraceptive depot medroxyprogesterone acetate (DMPA), with progestin as the single active compound, possesses selective glucocorticoid activity and can alter the expression of glucocorticoid receptor-regulated genes. We therefore propose that pharmacological doses of DMPA used for endocrine therapy could have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service