Skip to Content
Merck
All Photos(1)

Key Documents

33510

Sigma-Aldrich

1,10-Phenanthroline monohydrate

redox indicator, ACS reagent, puriss. p.a., ≥99.5% (calc. to the dried substance)

Synonym(s):

o-Phenanthroline monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2 · H2O
CAS Number:
Molecular Weight:
198.22
Beilstein:
4008096
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

product name

1,10-Phenanthroline monohydrate, ACS reagent, puriss. p.a., ≥99.5% (calc. to the dried substance), for redox titration

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.5% (calc. to the dried substance)
8.5-9.5% water basis

technique(s)

titration: suitable

impurities

8.5-9.5% water

ign. residue

≤0.05% (as SO4)

mp

100-104 °C (lit.)
97-102 °C

suitability

passes test for iron detection
passes test for redox indicator

SMILES string

O.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;/h1-8H;1H2

InChI key

PPQJCISYYXZCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,10-Phenanthroline monohydrate is the hydrated form of o-phenanthroline.

Application

1,10-Phenanthroline monohydrate (phen) may be used in the photochemical quantification of Fe3+ and total iron in mineral samples. It may be used as starting reagent for the preparation of 5-nitro-1,10-phenanthroline.
When complexed with copper, it possesses nuclease activity that has been used to study DNA-protein interactions.

Biochem/physiol Actions

Metalloprotease inhibitor; chelates iron, zinc and other divalent metals. Effective concentration 1-10 mM.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The quantitative assay of minerals for Fe2+ and Fe3+ using 1, 10-phenanthroline: II. A photochemical method.
Stucki JW.
Soil Science Society of America Journal. Soil Science Society of America, 45(3), 638-641 (1981)
Unmesh Jadhav et al.
Molecular cell, 78(1), 141-151 (2020-02-07)
Polycomb repressive complex 2 (PRC2) places H3K27me3 at developmental genes and is causally implicated in keeping bivalent genes silent. It is unclear if that silence requires minimum H3K27me3 levels and how the mark transmits faithfully across mammalian somatic cell generations.
A luminescent tris (2-thenoyltrifluoroacetonato) europium (III) complex covalently linked to a 1, 10-phenanthroline-functionalised sol-gel glass.
Binnemans K, et al.
Journal of Materials Chemistry, 14(2), 191-195 (2004)
Kyle R Simonetta et al.
Nature communications, 10(1), 1402-1402 (2019-03-31)
Protein-protein interactions (PPIs) governing the recognition of substrates by E3 ubiquitin ligases are critical to cellular function. There is significant therapeutic potential in the development of small molecules that modulate these interactions; however, rational design of small molecule enhancers of
Doo Sin Jo et al.
Autophagy, 16(11), 1989-2003 (2020-01-23)
Quality control of peroxisomes is essential for cellular homeostasis. However, the mechanism underlying pexophagy is largely unknown. In this study, we identified HSPA9 as a novel pexophagy regulator. Downregulation of HSPA9 increased macroautophagy/autophagy but decreased the number of peroxisomes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service