Skip to Content
Merck
All Photos(1)

Key Documents

31682

Supelco

Carpropamid

PESTANAL®, analytical standard

Synonym(s):

2,2-Dichloro-N-[1-(4-chlorophenyl)ethyl]-1-ethyl-3-methylcyclopropanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18Cl3NO
CAS Number:
Molecular Weight:
334.67
Beilstein:
8395957
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCC1(C(C)C1(Cl)Cl)C(=O)NC(C)c2ccc(Cl)cc2

InChI

1S/C15H18Cl3NO/c1-4-14(10(3)15(14,17)18)13(20)19-9(2)11-5-7-12(16)8-6-11/h5-10H,4H2,1-3H3,(H,19,20)

InChI key

RXDMAYSSBPYBFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carpropamid is widely used for the control of rice blast caused by Pyricularia oryzae. Carpropamid acts by inhibiting scytalone dehydratase enzyme involved in fungal melanin biosynthesis in a phytopathogenic fungus.

Application

Carpropamid may be used as a reference standard for the determination of the analyte in river water by liquid chromatography/time-of-flight mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tri Puji Priyatno et al.
TheScientificWorldJournal, 2012, 545784-545784 (2012-06-06)
The cyclic AMP- (cAMP-) dependent protein kinase A signaling pathway is one of the major signaling pathways responsible for regulation of the morphogenesis and pathogenesis of several pathogenic fungi. To evaluate the role of this pathway in the plant pathogenic
Carpropamid: a rice fungicide with two modes of action.
Thieron M, et al.
Pflanzenschutz-Nachrichten Bayer, 51(28), 257- 278 (1998)
A new on-line sample preparation system for the liquid chromatography/time-of-flight mass spectrometry simultaneous analysis of pesticides in river water.
Sasaki H, et al.
Analytical Sciences, 22(6), 835- 840 (2006)
Cryogenic X-ray crystal structure analysis for the complex of scytalone dehydratase of a rice blast fungus and its tight-binding inhibitor, carpropamid: the structural basis of tight-binding inhibition.
Nakasako M, et al.
Biochemistry, 37(28), 9931- 9939 (1998)
Young Seop Kwon et al.
Analytica chimica acta, 868, 60-66 (2015-03-31)
The sensitive detection of iprobenfos (IBF) and edifenphos (EDI) was successfully conducted by using a new aptamer-based colorimetric multi-aptasensor. The dissociation constants of this multi-target aptamer to both iprobenfos and edifenphos were found to be 1.67 μM and 38 nM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service