Skip to Content
Merck
All Photos(1)

Key Documents

31614

Supelco

Benzidine

analytical standard

Synonym(s):

4,4′-Diaminobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
Beilstein:
742770
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Nc1ccc(cc1)-c2ccc(N)cc2

InChI

1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2

InChI key

HFACYLZERDEVSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian J Pak et al.
PLoS neglected tropical diseases, 8(8), e3002-e3002 (2014-08-08)
Strongyloidiasis is a persistent human parasitic infection caused by the intestinal nematode, Strongyloides stercoralis. The parasite has a world-wide distribution, particularly in tropical and subtropical regions with poor sanitary conditions. Since individuals with strongyloidiasis are typically asymptomatic, the infection can
P D Josephy
Federation proceedings, 45(10), 2465-2470 (1986-09-01)
Benzidine oxidative activation may proceed by peroxidase-catalyzed one-electron oxidation via free radical intermediates, or by N-acetylation followed by monooxygenase-catalyzed N-hydroxylation. The peroxidase route has been examined by using horseradish peroxidase or prostaglandin H synthase in vitro. In the presence of
J Whysner et al.
Pharmacology & therapeutics, 71(1-2), 107-126 (1996-01-01)
The aromatic amine benzidine (BZ) has produced various tumors, including liver tumors, in mice, rats and hamsters. BZ forms DNA adducts in rodent liver, and it is positive in most genotoxicity tests. Only bladder tumors are produced in dogs and
T V Zenser et al.
Drug metabolism and disposition: the biological fate of chemicals, 26(9), 856-859 (1998-09-12)
Workers exposed to high levels of benzidine have a 100-fold increased incidence of bladder cancer. This review evaluates the overall metabolism of benzidine to determine pathways important to initiation of bladder cancer. Upon incubation of benzidine with liver slices from
H Akada et al.
Leukemia, 28(8), 1627-1635 (2014-02-01)
JAK2V617F is the most common mutation found in Philadelphia chromosome-negative myeloproliferative neoplasms (MPNs). Although a majority of MPN patients carry heterozygous JAK2V617F mutation, loss of heterozygosity (LOH) on chromosome 9p (9pLOH) involving the JAK2 locus has been observed in ∼30%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service