Skip to Content
Merck
All Photos(1)

Key Documents

28220

Supelco

Cumene

analytical standard

Synonym(s):

(1-Methylethyl)benzene, 2-Phenylpropane, Isopropylbenzene, NSC 8776

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)2
CAS Number:
Molecular Weight:
120.19
Beilstein:
1236613
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

8 mmHg ( 20 °C)
9.7 mmHg ( 37.7 °C)

Assay

≥99.5% (GC)

autoignition temp.

797 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.491 (lit.)
n20/D 1.491

bp

152-154 °C (lit.)

mp

−96 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1ccccc1

InChI

1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

RWGFKTVRMDUZSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cumene is a colourless aromatic liquid which is mainly used as solvent in production of phenol, acetone and acetophenone. It is insoluble in water and is miscible with organic solvents. It forms peroxide on exposure to air for long and reacts exothermically with concentrated nitric or sulphuric acid.

Application

It has been used as reference standard in GC-MS analysis for the determination of cumene in lake water sample.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Comprehensive Guide to the Hazardous Properties of Chemical Substances
A Comprehensive Guide to the Hazardous Properties of Chemical Substances, 1-523 (2007)
Determination of toxic toluene, xylene and cumene in different lake waters.
Hossain, Amzad M.; Kabir M. J. ; and Salehuddin, S. M.;
International Journal of Environmental Research and Public Health, 4.2, 341-346 (2010)
Johanna Bråred Christensson et al.
Contact dermatitis, 55(4), 230-237 (2006-09-09)
Concomitant positive reactions to colophonium, oxidized limonene, and/or oxidized linalool are recorded in patch test studies. The main allergens in these patch test mixtures are hydroperoxides, which form antigens by a radical pathway. Theoretically, concomitant reactions can be explained not
Mathieu Frenette et al.
Journal of the American Chemical Society, 128(51), 16432-16433 (2006-12-21)
A new class of thermally activated chain-breaking antioxidants is presented. Dimers of persistent carbon-centered radicals are able to inhibit the autoxidation of cumene and styrene with better rate constants than the commercial antioxidant Irganox HP-136 and 3,5-di-tert-butyl-4-hydroxyanisole. A dramatic increase
A Getsoian et al.
International journal of pharmaceutics, 348(1-2), 3-9 (2007-10-20)
To emphasize the fact that solvents can be either critical or immaterial in crystallizing specific polymorphs, a method for obtaining multiple polymorphs of a compound using only one solvent is demonstrated. By varying the crystallization temperature and level of supersaturation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service