Skip to Content
Merck
All Photos(1)

Documents

08748

Sigma-Aldrich

Triethylsulfonium bis(trifluoromethylsulfonyl)imide

for electrochemistry, ≥95.0% (qNMR)

Synonym(s):

[SEt3] NTf2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15F6NO4S3
CAS Number:
Molecular Weight:
399.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for electrochemistry

Quality Level

Assay

≥95.0% (qNMR)

form

liquid

impurities

≤200 ppm water

conductivity

~6.0 mS/cm

mp

-35.5 °C

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg

SMILES string

CC[S+](CC)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H15S.C2F6NO4S2/c1-4-7(5-2)6-3;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h4-6H2,1-3H3;/q+1;-1

InChI key

BLODSRKENWXTLO-UHFFFAOYSA-N

General description

Symmetric and aliphatic trialkylsulfonium cations form room temperature molten salts with bis(trifluoromethylsulfonyl)imide.

Application

Triethylsulfonium bis(trifluoromethylsulfonyl)imide is a room-temperature ionic liquid (RTIL ) that can be used:
  • As an electrolyte in electrochemical double-layer supercapacitors.
  • As a model ionic liquid used in the studies of structure effect of ions on electrochemical windows (EWs).
  • As a component of the gel polymer electrolyte, which is used in the construction of symmetrical electrical-double layer capacitors.
  • As an electrolyte in the in-plane supercapacitor devices.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanocomposites of graphene nanosheets/multiwalled carbon nanotubes as electrodes for in-plane supercapacitors
Trigueiro JPC, et al.
Electrochimica Acta, 187(1), 312-322 (2016)
Application of novel room temperature ionic liquids in flexible supercapacitors
Wei Di and Ng TW
Electrochemical Communications, 11(10), 1996-1999 (2009)
Activated graphene oxide/reduced graphene oxide electrodes and low viscous sulfonium cation based ionic liquid incorporated flexible gel polymer electrolyte for high rate supercapacitors
Suleman M, et al.
Journal of alloys and compounds, 695(1), 3376-3392 (2017)
Matsumoto, H., et al.
Chemistry Letters (Jpn), 29, 1430-1430 (2000)
Investigating the electrochemical windows of ionic liquids.
Hayyan M, et al.
Journal of Industrial and Engineering Chemistry, 19(1), 106-112 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service