Skip to Content
Merck
All Photos(2)

Key Documents

06808

Supelco

(+)-Valencene

analytical standard

Synonym(s):

(3R,4aS,5R)-4a,5-Dimethyl-3-isopropenyl-1,2,3,4,4a,5,6,7-octahydronaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
3539153
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥80.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.504 (lit.)

bp

274 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1CCC=C2CC[C@H](C[C@@]12C)C(C)=C

InChI

1S/C15H24/c1-11(2)13-8-9-14-7-5-6-12(3)15(14,4)10-13/h7,12-13H,1,5-6,8-10H2,2-4H3/t12-,13-,15+/m1/s1

InChI key

QEBNYNLSCGVZOH-NFAWXSAZSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Stepwise engineering of Saccharomyces cerevisiae to produce (+)-valencene and its related sesquiterpenes: Researchers developed a method to enhance the production of (+)-valencene and other sesquiterpenes in yeast by systematic genetic modifications, suggesting applications in bio-manufacturing of these compounds (Ouyang X et al., 2019).

  • Production of glutamate and stereospecific flavors, (S)-linalool and (+)-valencene, by Synechocystis sp. PCC6803: This article presents a novel approach using the cyanobacterium Synechocystis sp. for the production of both glutamate and specific terpene flavors, demonstrating the organism′s potential in biotechnological applications (Matsudaira A et al., 2020).

  • Production, Function, and Applications of the Sesquiterpenes Valencene and Nootkatone: a Comprehensive Review: This comprehensive review discusses the biochemical pathways, production methods, and commercial applications of valencene and nootkatone, highlighting their significance in the pharmaceutical and flavor industries (Zhang LL et al., 2023).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service