Skip to Content
Merck
All Photos(1)

Key Documents

05290

Millipore

Sterility Indicator (Radiation Sterilization)

suitable for microbiology

Synonym(s):

Spore Strips (for Radiation Sterilization), Spore Strips Bioindicator (for Radiation Sterilization)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

shelf life

limited shelf life, expiry date on the label

application(s)

clinical
cosmetics
environmental
food and beverages
pharmaceutical
veterinary

microbiology

storage temp.

2-8°C

General description

Sterility Indicator (Radiation Sterilization) consists of Bacillus pumilus (ATCC 27142) spores impregnated on paper strips, individually placed into envelopes. The number of spores present per strip: 1 million. Bacillus pumilus is a radiation resistance species. The spores are highly resistant and are used to monitor the efficiency of radiation sterilization. These indicators as specified by U.S. military specification MIL-S-36586 are GMP requirements of U.S. FDA.

Packaging

package of 25 strips
Dimension envelop (2 strips): 46x70 mm
Dimension strips: 33x7 mm

Components

Bacillus pumilus (ATCC 27142) spores impregnated on paper strips (1 mio. spores per strip)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endospores as biological indicator for the validation of sterilization process
CHEUNG H Y
Pharmaceutical Technology, 158-158 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service