Skip to Content
Merck
All Photos(1)

Key Documents

04163

Millipore

Nutrient Agar No 2, Vegitone

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Vegitone Nutrient Agar No 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

Agency

according to ISO 6579-1:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
peptone (vegetable), 5.0 g/L
vegetable extract, 3.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
life science and biopharma
water monitoring

microbiology

suitability

nonselective for bacteria (General Media)

Application

This Nutrient Agar contains only plant peptone instead of animal peptone. It is used for the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces and other materials.

Preparation Note

Suspend 23 g in 1000 ml distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. If desired, the medium can be enriched with 5-10% v/v sterile defibrinated blood.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service