Skip to Content
Merck
All Photos(1)

Key Documents

01495

Supelco

L-Glutamic acid monosodium salt hydrate

analytical standard

Synonym(s):

(S)-2-Aminopentanedioic acid, L-Glutamic acid monosodium salt, Glu, MSG, Monosodium glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8NNaO4 · xH2O
CAS Number:
Molecular Weight:
169.11 (anhydrous basis)
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E620
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥98.5% (TLC)

shelf life

limited shelf life, expiry date on the label

impurities

9.62% water (theory, monohydrate)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

InChI

1S/C5H9NO4.Na.H2O.H/c6-3(5(9)10)1-2-4(7)8;;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2;

InChI key

JYDGDQPVBSBYRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monosodium L-glutamate-induced asthma.
Allen, David H., John Delohery, and Gary Baker.
The Journal of Allergy and Clinical Immunology, 80.4, 530-537 (1987)
E von Hauff et al.
Biosensors & bioelectronics, 26(2), 596-601 (2010-08-07)
A biocompatible device for the voltage dependent uptake and release of the neural transmitter L-glutamate in neutral pH solutions is demonstrated. The device consists of a gold electrode coated with molecularly imprinted, overoxidised polypyrrole (oPPy). It is shown here that
Reza Rezaei et al.
Amino acids, 44(3), 911-923 (2012-11-03)
Dietary intake of glutamate by postweaning pigs is markedly reduced due to low feed consumption. This study was conducted to determine the safety and efficacy of dietary supplementation with monosodium glutamate (MSG) in postweaning pigs. Piglets were weaned at 21
H Hosaka et al.
Alimentary pharmacology & therapeutics, 36(9), 895-903 (2012-09-18)
Monosodium l-glutamate (MSG) is known to influence the endocrine system and gastrointestinal (GI) motility. The mechanism of postprandial glycemic control by food in the GI tract is mostly unknown and of great interest. To investigate the effect of MSG on
O A Rotimi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(11), 4062-4067 (2012-08-18)
In order to investigate the influence of some fibre-enriched diets on tissue lipids in an animal model of obesity induced by the administration of monosodium glutamate (MSG), obese rats were fed diets containing 30% of Acha, Cassava, Maize and Plantain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service