Skip to Content
Merck
All Photos(1)

Key Documents

01388

Supelco

(−)-Gallocatechin

analytical standard

Synonym(s):

(2S,3R)-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@H]1c3cc(O)c(O)c(O)c3

InChI

1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15+/m1/s1

InChI key

XMOCLSLCDHWDHP-DOMZBBRYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gallocatechin belongs to the catechin class of organic compounds and is one of the major components of green tea. The beneficial effects of catechins include anti-tumorigenic, anti-mutagenic, anti-pathogenic, and anti-oxidative properties.

Application

(−)-Gallocatechin may be used as an analytical standard for the determination of the analyte in biological fluids and tea beverages by chromatography-based techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of catechins and catechin gallates in biological fluids by HPLC with coulometric array detection and solid phase extraction
Chu KO, et al.
Analytica Chimica Acta, 510(1), 69-76 (2004)
Joaudimir Castro et al.
Talanta, 82(5), 1687-1695 (2010-09-30)
Presented here is the quantitative analysis of green tea NIST standard reference materials (SRMs) via liquid chromatography-particle beam/electron ionization mass spectrometry (LC-PB/EIMS). Three different NIST green tea standard reference materials (SRM 3254 Camellia sinesis Leaves, SRM 3255 C. sinesis Extract
Determination of catechins in human plasma after commercial canned green tea ingestion by high-performance liquid chromatography with electrochemical detection using a microbore column
Kotani A, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 788(2), 269-275 (2003)
Hongyu Wang et al.
Journal of agricultural and food chemistry, 60(12), 3098-3104 (2012-03-08)
Proanthocyanidins in unripe Manilkara zapota (chiku) were isolated using solvent extraction followed by Sephadex LH-20 fractionation with a yield of 0.9%. HPLC analysis using a diol column revealed well-resolved oligomers ranging from dimer to hexamer. The majority of the proanthocyanidins
Yoo-Hyun Lee et al.
International journal of molecular medicine, 30(1), 69-74 (2012-04-17)
Manipulating acetylation status of key gene targets is likely to be crucial for effective cancer therapy. In this study, we utilized green tea catechins, epicatechin (EC), epigallocatechin (EGC) and epigallocatechin-3-gallate (EGCG) to examine the regulation of androgen receptor acetylation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service