Skip to Content
Merck
All Photos(2)

Documents

SCR006

Sigma-Aldrich

AccumaxCell Counting Solution in DPBS Sterile-filtered Cell Culture Tested

A cell detachment solution of proteolytic, collagenolytic & DNAse enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.

Synonym(s):

Accumax Cell Solution, Cell Counting Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160801
NACRES:
NA.78

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

input

sample type mesenchymal stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type epithelial cells
sample type cardiac stem cell(s)
sample type: human embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type neural stem cell(s)

shipped in

dry ice

General description

A cell detachment solution of proteolytic, collagenolytic and DNAse enzymes. Useful on a variety of cells lines however not all cells or cell types are affected by ACCUMAX, also passage number and confluency can influence how well the reagent does because the amounts and makeup of the collagen extracellular matrix will vary with age and culture state. Because ACCUMAX is a more gentle, less univerally proteolytic cleavage mixture reagent amounts and incubation times need to be optimized for each cell system.

The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue and the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including SmartPlastic. ACCUMAX does not contain mammalian or bacterial derived products.

Each lot of ACCUMAX is tested for Sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic.



ACCUMAX has been used successfully on numerous cell types including: CHO, BHK, 293, COS, Sf9, and catfish primary hepatocyte cells.

Application

A cell detachment solution of proteolytic, collagenolytic & DNAse enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.

Physical form

100 ml, ready to use, frozen sterile liquid. 1X ACCUMAX enzymes in Dulbecco′s PBS (0.2 g/L KCl, 0.2 g/L KH2PO4, 8 g/L NaCl, and 1.15 g/L Na2HPO4) containing 0.5mM EDTA-4Na.

Storage and Stability

Stable when stored at -20°C. Refer to lot expiration date. Recommended storage upon receipt is -20°C. After thawing, ACCUMAX may be stored for up to 2 months at 4°C. DO NOT STORE AT ROOM TEMP. Use sterile technique when handling.

Legal Information

Accumax is a trademark of Innovative Cell Technologies, Inc.
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jacqueline S Penaloza et al.
Biochemical and biophysical research communications, 520(2), 284-290 (2019-10-09)
The Mesp1 lineage contributes to cardiac, hematopoietic and skeletal myogenic development. Interestingly, muscle stem cells residing in craniofacial skeletal muscles primarily arise from Mesp1+ progenitors, but those in trunk and limb skeletal muscles do not. To gain insights into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service