Skip to Content
Merck
All Photos(1)

Key Documents

QIA97

Sigma-Aldrich

Cu/Zn Superoxide Dismutase ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

usage

sufficient for 96 tests

Quality Level

species reactivity

human

manufacturer/tradename

Calbiochem®

packaging

pkg of  96-well plate(s)

storage condition

OK to freeze

assay range

sensitivity: 40 pg/mL

technique(s)

ELISA: suitable

input

plasma
serum
urine

detection method

colorimetric

shipped in

wet ice

storage temp.

2-8°C

General description

A colorimetric sandwich ELISA method for detecting Cu/Zn superoxide dismutase (SOD) in different types of biological samples.

Packaging

1 kit in glass bottle

Components

Pre-Coated 96-Well Plate, Anti-Cu/Zn SOD/HRP-Conjugate, Cu/Zn SOD Standard, Wash Buffer Concentrate, Assay Buffer Concentrate, Phosphate Buffered Saline, Substrate Solution, Stop Solution, Plate Covers, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Specifications

Assay time: 1.5 h

Storage and Stability

Upon arrival store the entire contents of the kit at 4°C.

Other Notes

Herouart, D., et al. 1993. Proc. Natl. Acad. Sci. USA 90, 3108.
Niwa, Y., et al. 1993. Am. J. Pathol. 143, 312.
Suzuki, H., et al. 1993. J. Clin. Invest. 91, 2727.
Holzgreve, W., et al. 1991. Diagnose Labor. 41, 162.
Porstmann, T., et al. 1991. Prenat. Diagn. 11, 195.
Shull, S., et al. 1991. J. Biol. Chem. 266, 24398.
Porstmann, T., et al. 1990. Hum. Genet. 85, 362.
Porstmann, T., et al. 1988. Clin. Chim. Acta 171, 1.
Jolly, S.R., et al. 1984. Circ. Res. 54, 277.
Goebel, K.M. and Storck, U. 1983. Am. J. Med. 74, 124.
Barra, D., et al. 1980. FEBS Lett. 120, 53.
Sinet, P.M., et al. 1976. Exp. Cell Res. 97, 47.
Harta, J.W. and Deutsch, H.F. 1972. Biol. Chem. 247, 7043.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The Calbiochem® Cu/Zn Superoxide Dismutase ELISA Kit is designed to measure Cu/Zn superoxide dismutase (SOD) in cell culture supernatants and biological fluids like human serum, plasma, urine, and amniotic fluid.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service