Skip to Content
Merck
All Photos(1)

Key Documents

GSS-1015

Sigma-Aldrich

GS System 100mM L-Methionine sulfoximine, working range 25µM to 1mM

The GS System (offered by & a registered trademark of Celltech Biologicals, Inc.) utilizes specific vectors for cell culture transfection.

Synonym(s):

100mM LMSX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

manufacturer/tradename

Specialty Media

General description

The GS System products are developed with the highest quality reagents and ultra-pure H2O. It employs specific vectors for cell culture transfection. Specialty Media provides media and reagents for use with this system.

Application

GS System 100mM L-Methionine sulfoximine, working range 25μM to 1mM has been used as a component in CD Chinese hamster ovary (CHO) medium to resuspend freshly thawed CHO-K1SP cells.

Legal Information

GS System is a trademark of Lonza Group Ltd.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corinna Lau et al.
mAbs, 12(1), 1686319-1686319 (2019-11-02)
The mechanism of action of recombinant IgG2/4 antibodies involves blocking of their target without the induction of effector functions. Examples are eculizumab (Soliris®), which is used clinically to block complement factor C5, as well as anti-human CD14 (r18D11) and anti-porcine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service