Skip to Content
Merck
All Photos(1)

Documents

CBL186

Sigma-Aldrich

Anti-DNA Antibody, clone AC-30-10

clone AC-30-10, Chemicon®, from mouse

Synonym(s):

Deoxyribonucleic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AC-30-10, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgM

shipped in

wet ice

target post-translational modification

unmodified

Specificity

This antibody reacts with native denatured DNA extracted from various organisms: mouse DNA, calf thymus DNA, M13 DNA, SV-40 DNA and several plasmids. It also reacts with synthetic DNA of different base compositions: poly dAT-poly dAT, poly dAT. DNA-containing structures such as chromatin in diverse tissues from several species as well as in diverse cell lines such as XLKE-A6 (Xenopus), RVF-SMC (rat kangaroo) and HeLa (human) are also recognized.

Application

Anti-DNA Antibody, clone AC-30-10 is a Mouse Monoclonal Antibody for the detection of DNA & has been validated in IF, IHC(P).
Research (cell biology, molecular biology, microbiology) since it reacts with any form of native and denatured DNA. Reliable positive control in immunofluoresence assays for detecting DNA antibodies on tissues of cultured cells, notably in the diagnosis of autoantibodies and autoimmune diseases.

Sensitive detection of mycoplasma contamination in cell cultures.
For frozen and paraffin tissue sections.

Optimal working dilutions must be determined by the end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Physical form

Format: Purified
The immunoglobulin was purified by gel filtration and presented in PBS, pH 7.4, containing 0.09% sodium azide and 0.5% BSA.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andong Qin et al.
Cells, 12(21) (2023-11-10)
Coxsackievirus B3 (CVB3), a single-stranded positive RNA virus, primarily infects cardiac myocytes and is a major causative pathogen for viral myocarditis (VMC), driving cardiac inflammation and organ dysfunction. However, whether and how myocardial damage is involved in CVB3-induced VMC remains
Sebastian Aguirre et al.
Nature microbiology, 2, 17037-17037 (2017-03-28)
During the last few decades, the global incidence of dengue virus (DENV) has increased dramatically, and it is now endemic in more than 100 countries. To establish a productive infection in humans, DENV uses different strategies to inhibit or avoid
Pedro Silva-Pinheiro et al.
Nucleic acids research, 49(9), 5230-5248 (2021-05-07)
Mutations in POLG, encoding POLγA, the catalytic subunit of the mitochondrial DNA polymerase, cause a spectrum of disorders characterized by mtDNA instability. However, the molecular pathogenesis of POLG-related diseases is poorly understood and efficient treatments are missing. Here, we generate
Genome-wide analysis reveals coating of the mitochondrial genome by TFAM.
Wang, YE; Marinov, GK; Wold, BJ; Chan, DC
Testing null
Mohammad T Hussan et al.
Current issues in molecular biology, 44(3), 1215-1223 (2022-06-21)
The presence of ectopic DNA in the cytoplasm induces inflammation and cell death. It has been widely reported that leakage of nuclear DNA into the cytoplasm can mainly be sensed by cyclic GMP-AMP synthase (cGAS). We recently reported that mitochondria-derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service