Skip to Content
Merck
All Photos(1)

Key Documents

AP192A

Sigma-Aldrich

Donkey Anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed

0.6 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

0.6 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Mouse IgG (H+L) (absorbed for minimal cross-reactivity to Bovine, Chicken, Goat, Guinea Pig, Syrian Hamster, Horse, Human, Sheep, Rabbit, and Rat Serum Proteins)

Application

ELISA and Western Blotting at 1:5000 - 1:50,000 dilution.

Optimal working dilutions must be determined by the end user.
This Donkey anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed is validated for use in ELISA, WB for the detection of Donkey Mouse IgG.

Physical form

Lyophilized. Buffer = 0.01M Tris-HCl, 0.25M NaCl, pH 8.0, with 15mg/mL BSA and 0.05% Sodium Azide.

RECONSTITUTION:

Reconstitute to 1.0 mg/mL with 0.5 mL sterile distilled water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service