Skip to Content
Merck
All Photos(1)

Documents

AG56P

Sigma-Aldrich

Human Laminin (pepsinized) Purified Protein

Synonym(s):

Laminin 1, Laminin 2, Laminin 3, Laminin 6, Laminin 8, Laminin 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

biological source

human

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Chemicon®

concentration

0.25 mg/mL

technique(s)

cell culture | mammalian: suitable

input

sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type epithelial cells
sample type: human embryonic stem cell(s)
sample type mesenchymal stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C
−70°C

Gene Information

human ... LAMB1(3912)

General description

AG56P contains a mixture of human laminins containing the beta1 chain, principally laminin 1, 2, 3, 6, 8 & 10. This preparation is immunologically and biologically identical to intact human laminin. AG56P is suitable for use in ELISA, production of antiserum, cell adhesion and attachment assays, and neurite-stimulation assays. This pepsinized laminin preparation runs as twin bands migrating at approximately 160kDa and 130kDa by SDS-PAGE under reducing conditions.

Preparation: Chemicon′s purified human laminin is prepared from freshly frozen human placenta tissue that is homogenized in PBS with a blender and the mixture centrifuged. The pellet is then washed with 0.5M acetic acid, collected and subjected to and extended Pepsin A digestion. The pepsin digest is neutralized, centrifuged to clear debris, and applied to an antibody column containing mouse anti-human laminin monoclonal 4E10 monoclonal antibody {MAB1921}. The solubilized laminin solution is passed over the column, and utlimately the laminin is released via acidification with KSCN. Then the material is dialyzed against PBS. The antibody 4E10 is specific for laminin beta1 chain. This chain is found in Laminin′s 1,2,3,6,8,10 under the native conditions used to purify the material.
Product Source: Human placenta, tested negative for hepatitis B virus, hepatitis C virus (HCV), HIV-1, HIV-2, HTLV-1, HTLV-2, and Treponema pallidum. Handle as if potentially infectious.

Quality

Each lot is analyzed on SDS-PAGE for band integrity and expected sizes prior to packaging and release

Physical form

Purified laminin protein in liquid in Tris Buffer Saline with 0.01% sodium azide.

Preparation Note

Purified from a mild pepsin digestion followed by affinity chromatography on monoclonal (4E10) anti-human laminin-Sepharose.

Storage and Stability

Store at -20°C or -70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a table-top centrifuge to dislodge any liquid in the container′s cap.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Zaslavsky et al.
FEBS letters, 579(18), 3899-3906 (2005-07-01)
Several different types of interactions between sphingosine-1-phosphate (S1P) receptors and platelet-derived growth factor receptor (PDGFR) have been revealed recently. In this work, we used HEK293 cells to further investigate the potential crosstalk. Interestingly, we observed that S1P specifically induced a
Mapping of domains in human laminin using monoclonal antibodies: localization of the neurite-promoting site.
Engvall, E, et al.
The Journal of cell biology, 103, 2457-2465 (1986)
Jan-Eric Ahlfors et al.
Stem cell research & therapy, 10(1), 166-166 (2019-06-15)
Cell reprogramming is a promising avenue for cell-based therapies as it allows for the generation of multipotent, unipotent, or mature somatic cells without going through a pluripotent state. While the use of autologous cells is considered ideal, key challenges for
Domenico Vitolo et al.
The American journal of pathology, 168(3), 991-1003 (2006-03-02)
Capillaries expressing the laminin alpha2 chain in basement membranes may be considered early developing vessels in normal and neoplastic human tissues. Therefore, we investigated whether up-regulation of this extracellular matrix protein favors transendothelial migration of neoplastic cells and then metastasis.
Colin D Paul et al.
Biomaterials, 197, 101-118 (2019-01-15)
Biophysical aspects of in vivo tissue microenvironments include microscale mechanical properties, fibrillar alignment, and architecture or topography of the extracellular matrix (ECM). These aspects act in concert with chemical signals from a myriad of diverse ECM proteins to provide cues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service