Skip to Content
Merck
All Photos(2)

Documents

ABE483

Sigma-Aldrich

Anti-TFAM Antibody

serum, from rabbit

Synonym(s):

Transcription factor A, mitochondrial, mtTFA, Testis-specific high mobility group protein, TS-HMG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TFAM(7019)
mouse ... Tfam(21780)

General description

TFAM, also called mtTFA, is a mitochondrial transcription factor that is a key activator of mitochondrial transcription as well as a participant in mitochondrial genome replication. Studies in mice have demonstrated that this gene product is required to regulate the mitochondrial genome copy number and is essential for embryonic development.

Immunogen

His-tagged recombinant protein corresponding to mouse mtTFA.

Application

Detect TFAM using this rabbit polyclonal antibody, Anti-TFAM Antibody validated for use in western blotting & IP.
Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated TFAM in C2C12 cell lysate (Prof. B Kaufman, University of Pennsylvania.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TFAM in 10 µg of C2C12 cell lysate.

Target description

~27 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-Hyuk Lee et al.
Experimental & molecular medicine, 54(12), 2135-2147 (2022-12-07)
PARPs play fundamental roles in multiple DNA damage recognition and repair pathways. Persistent nuclear PARP activation causes cellular NAD+ depletion and exacerbates cellular aging. However, very little is known about mitochondrial PARP (mtPARP) and poly ADP-ribosylation (PARylation). The existence of
Nicole DeFoor et al.
Scientific reports, 13(1), 15339-15339 (2023-09-16)
SARS-CoV-2 causes the severe respiratory disease COVID-19. Remdesivir (RDV) was the first fast-tracked FDA approved treatment drug for COVID-19. RDV acts as an antiviral ribonucleoside (adenosine) analogue that becomes active once it accumulates intracellularly. It then diffuses into the host
Wenci Weng et al.
Experimental and therapeutic medicine, 23(3), 239-239 (2022-03-01)
Type 1 diabetes (T1D) is characterized by dysregulated blood glucose and liver metabolism. In previous studies, niclosamide ethanolamine salt (NEN) and artemether (Art) displayed significant hypoglycemic effects. However, their combined therapeutic effects on the liver in T1D have remained elusive.
Krishna Prahlad Maremanda et al.
Toxicology and applied pharmacology, 385, 114788-114788 (2019-11-05)
Cigarette smoke (CS)-induced lung inflammation and Chronic Obstructive Pulmonary disease (COPD) involves mitochondrial dysfunction. Mesenchymal stem cells (MSC) and MSC-derived exosomes (EXO) are reported to show therapeutic effects in many animal models of inflammation and injury. In the present study
Sylvia Torres-Odio et al.
Journal of immunology (Baltimore, Md. : 1950), 206(8), 1890-1900 (2021-03-19)
Caseinolytic mitochondrial matrix peptidase proteolytic subunit (CLPP) is a serine protease that degrades damaged or misfolded mitochondrial proteins. CLPP-null mice exhibit growth retardation, deafness, and sterility, resembling human Perrault syndrome, but also display immune system alterations. However, the molecular mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service