Skip to Content
Merck
All Photos(1)

Key Documents

48-610MAG

Millipore

MILLIPLEX MAP STAT Cell Signaling Magnetic Bead 5-Plex Kit - Cell Signaling Multiplex Assay

Synonym(s):

Magnetic Bead STAT Kit, STAT Cell Signaling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

storage temp.

2-8°C

General description

The STAT (Signal Transducers and Activators of Transcription) family, consisting of seven mammalian members, regulates many aspects of growth, survival and differentiation in cells. The extracellular binding of cytokines, hormones, growth factors and neurotransmitters to receptors induces phosphorylation of the intracellular Janus kinases (JAKs) that, in turn, phosphorylate STAT proteins at conserved tyrosine residues, activating SH2-mediated dimerization followed by translocation to the nucleus. STAT dimers then bind to IRE (interferon response element) and GAS (gamma interferon-activated sequence) DNA elements, resulting in the transcriptional regulation of downstream genes.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • An overnight (4°C) incubation is recommended for best results.
  • This assay requires 25 μL diluted cell lysate per well.
  • This kit must be run using Assay Buffer 1 (provided).
  • 1 - 25 μg cell lysate/well (recommended starting concentration is 40 to 1,000 μg protein/mL).

Research Category
Signaling|Apoptosis & Cancer
The analytes available for this multiplex kit are: STAT1 (Tyr701), STAT2 (Tyr690), STAT3 (Tyr705), STAT5A/B (Tyr694/699), STAT6 (Tyr641).
Used to detect/quantify:STAT1 (Tyr701)|STAT2 (Tyr690)|STAT3 (Tyr705)|STAT5A/B (Tyr694/699)|STAT6 (Tyr641)

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service