Skip to Content
Merck
All Photos(1)

Key Documents

407303

Sigma-Aldrich

γ-Interferon, Mouse, Recombinant, E. coli

Synonym(s):

IFNG, Type II IFN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

solubility

water: soluble

Protein ID accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

mouse ... Ifng(15978)

General description

Note: 1 KU = 1000 units.
Research area: Immunology and Cytokines

Recombinant, mouse γ-interferon, consisting of 134 amino acids, expressed in E. coli as a single, non-glycosylated polypeptide.

Application

γ-Interferon has been used to assess the response of wild-type and cytosolic phospholipase A2 (cPLA2)-deficient cells to IFN-mediated signals. It has also been used to study the signaling pathway associated with IFN-γ and to explore the potential role of β-Arrestin1 in the dephosphorylation of signal transducer and activator of transcription 1 (STAT1).

Biochem/physiol Actions

Interferon-gamma (IFN-γ), a proinflammatory cytokine, exhibits a significant impact on innate and adaptive immunity in terms of combatting intracellular infections and facilitating tumor control. It has a significant impact on various immunological pathways. IFN-γ influences the following biological processes:

Antigen Processing and Presentation: IFN-γ enhances the expression of molecules involved in antigen presentation. This helps in the efficient activation of T cells, leading to the initiation of adaptive immunity.

Reactive Oxygen Species (ROS): IFN-γ stimulates the production of ROS by promoting the assembly of NADPH oxidase subunits. These ROS possess strong microbicidal effects, aiding in the defense against invading pathogens.

Autophagy: IFN-γ induces the expression of proteins involved in autophagy, facilitating the clearance of intracellular pathogens and contributing to immune defense.

Apoptosis: IFN-γ triggers apoptotic cell death by increasing the expression of molecules associated with the apoptotic pathway.

Cell Migration and Leukocyte Traffic: IFN-γ promotes the expression of chemokines and adhesion molecules, facilitating immune cell migration and contributing to the initiation of inflammatory responses.

Antiviral Mediators: By inducing the expression of various antiviral proteins, IFN-γ counteracts viral infections at multiple stages of the viral life cycle.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, pH 7.4.

Preparation Note

Dilute only in buffers containing protein such as 0.1% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Zantl, N., et al. 1998. Infect. Immun. 66, 2300.
Familletti, P.C., et al. 1981. Methods Enzymol. 78, 387.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

cPLA2 regulates the expression of type I interferons and intracellular immunity to Chlamydia trachomatis
Vignola MJ, et al.
The Journal of Biological Chemistry, 285(28), 1625-1635 (2010)
Impact of interferon-? on hematopoiesis
Bruin AMD, et al.
Blood, 24(16), 2479-2486 (2014)
Interferon-gamma (IFN-?): Exploring its implications in infectious diseases
Kak G, et al.
Biomolecular Concepts, 64-79, 9(1)-9(1) (2018)
Pei-Ju Liu et al.
Journal of the American Society of Nephrology : JASN, 33(11), 1989-2007 (2022-11-01)
Myo1e is a nonmuscle motor protein enriched in podocytes. Mutations in <i>MYO1E</i> are associated with steroid-resistant nephrotic syndrome (SRNS). Most of the <i>MYO1E</i> variants identified by genomic sequencing have not been functionally characterized. Here, we set out to analyze two
Christin Pelzel et al.
Molecular cell, 50(1), 149-156 (2013-04-16)
Signal transducer and activator of transcription 1 (STAT1) is activated by tyrosine phosphorylation upon interferon-γ (IFNγ) stimulation, which results in the expression of genes with antiproliferative and immunomodulatory functions. The inactivation of STAT1 occurs through tyrosine dephosphorylation by the tyrosine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service