1.41350
Polyvinyl alcohol
4-88, EMPROVE® ESSENTIAL, Ph. Eur., ChP, JPE, USP
Pharma Manufacturing
Synonym(s):
Poly(vinyl alcohol), PVA, PVOH
About This Item
Recommended Products
Agency
ChP
JPE
Ph. Eur.
USP
Quality Level
product line
EMPROVE® ESSENTIAL
form
scales
technique(s)
API processing | nano-milling: suitable
mp
160-240 °C
application(s)
controlled release
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
15-25°C
InChI
1S/C2H4O/c1-2-3/h2-3H,1H2
InChI key
IMROMDMJAWUWLK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service