Skip to Content
Merck
All Photos(1)

Documents

1.19778

Supelco

Calcium standard solution

traceable to SRM from NIST Ca(NO₃)₂ in HNO₃ 0.5 mol/l 1000 mg/l Ca Certipur®

Synonym(s):

Calcium reference standard solution for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7

product line

Certipur®

concentration

1000 mg/L Ca in nitric acid

technique(s)

AAS: suitable

pH

0.5 (20 °C in H2O)

density

1.014 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The calcium standard solution may find its following use:
  • To determine metals in milk samples using flame atomic absorption spectroscopy (FAAS) and complexometric titration with EDTA
  • Estimation of calcium in grape must and base wine by fourier transform near-infrared (FT-NIR) spectroscopy
  • In the method development to determine calcium, magnesium, potassium, iron, copper, zinc, manganese and phosphorus in bread samples by microwave plasma-atomic emission spectrometry (MIP-AES)
  • Validation of an alternative method to measure calcium and magnesium in biological fluids
  • Determination of calcium and zinc in gluconate oral solutions and blood samples by liquid cathode glow discharge-atomic emission spectroscopy (LCGD-AES)
  • Development and validation of an inductively coupled plasma-atomic emission spectroscopy (ICP-AES) based method to determine trace elements in vinegar

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization and validation of candidate reference methods for the determination of calcium and magnesium in biological fluids
Aljerf L and Mashlah A
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 132, 411-421 (2017)
Development and Validation of an Inductively Coupled Plasma-Atomic Emission Spectrometry (ICP-AES) Method for Trace Element Determination in Vinegar
Paktsevanidou IP, et al.
Analytical Letters, 54(13), 2227-2238 (2021)
Comparative study for the determination of metals in milk samples using flame-AAS and EDTA complexometric titration
Hussain Z, et al.
Journal of science, technology and environment, 40(1), 1-6 (2010)
Determination of calcium and zinc in gluconates oral solution and blood samples by liquid cathode glow discharge-atomic emission spectrometry
Yu J, et al.
Talanta, 175, 150-157 (2017)
Predicting calcium in grape must and base wine by FT-NIR spectroscopy
Vestia J, et al.
Food Chemistry, 276, 71-76 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service