Skip to Content
Merck

1.03583

SAFC

Parteck® SI 150 (Sorbitol)

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF, JSFA, E 420

Pharma Manufacturing

Synonym(s):

Parteck® Sorbitol, D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352201
E Number:
E420

Agency

BP
JP
JSFA
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

fine powder

autoignition temp.

420 °C

particle size

(see specification)

pH

6-7 (20 °C, 100 g/L in H2O)

mp

94-96 °C

transition temp

flash point >100 °C

solubility

2350 g/L

density

1.49 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-25°C

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2

InChI key

FBPFZTCFMRRESA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® SI excipient is a directly compressible sorbitol which combines excellent tableting behavior, resulting in very robust and shiny tablets, with a pleasant mouthfeel and taste. The unique and large surface area allows for a fast disintegration and dissolution, even at high tablet hardnesses and supports a good content uniformity of low dose API formulations. Parteck® SI excipient comes in a variety of particle sizes and qualities to meet the needs of a very wide range of applications.

Features and Benefits

  • Excellent direct compression properties
  • High dilution and adsorption capacity
  • Larger and highly structured surface area
  • Low hygroscopicity
  • Low amount of reducing sugars

Storage and Stability

Storage temperature: no restrictions.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service