Skip to Content
Merck
All Photos(1)

Key Documents

1.00582

Supelco

Tungstophosphoric acid hydrate

cryst. EMPLURA®

Synonym(s):

Tungstophosphoric acid hydrate, Phosphotungstic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(W3O10)4] · x H2O
CAS Number:
Molecular Weight:
2880.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352301
EC Index Number:
215-682-7
NACRES:
NA.21

Quality Level

product line

EMPLURA®

form

solid

potency

3300 mg/kg LD50, oral (Rat)

loss

≤17% loss on ignition, 750 °C

pH

1.3 (20 °C, 20 g/L in H2O)

mp

107 °C

solubility

water: soluble at 20 °C

bulk density

960 kg/m3

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Cu: ≤0.001%
Fe: ≤0.002%
Pb: ≤0.005%

storage temp.

15-25°C

InChI

1S/H3O4P.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

AVFBYUADVDVJQL-UHFFFAOYSA-N

Analysis Note

Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.01 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.002 %
Pb (Lead): ≤ 0.005 %
Loss on ignition (750 °C): ≤ 17 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service