Skip to Content
Merck
All Photos(1)

Key Documents

07-383

Sigma-Aldrich

Anti-Akt3/PKBγ Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AKT3(10000)

Specificity

Recognizes Akt3/PKBγ.

Immunogen

peptide corresponding to amino acids 119-131 of human Akt3/PKBgamma

Application

Anti-Akt3/PKBγ Antibody is an antibody against Akt3/PKBγ for use in IP & WB.

Quality

routinely evaluated by immunoprecipitating active Akt3/PKBγ from IGF-I stimulated HEK293 cell lysates.

Target description

~60 kDa

Linkage

Replaces: AB3136

Physical form

Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.

Analysis Note

Control
IGF-I stimulated HEK293 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Joy et al.
Journal of neuro-oncology, 130(1), 43-52 (2016-10-21)
The growth factor receptor/PI3K/AKT pathway is an important drug target in many cancers including Glioblastoma. AKT, a key node in the pathway, has 3 isoforms, AKT1, AKT2 and AKT3. Here we investigate their role in GBM. We find each activated
Conrad P Hodgkinson et al.
Biochemistry, 41(32), 10351-10359 (2002-08-07)
Protein kinase B (PKB), also known as Akt, is a serine/threonine protein kinase controlled by insulin, various growth factors, and phosphatidylinositol 3-kinase. Full activation of the PKB enzyme requires phosphorylation of a threonine in the activation loop and a serine
François Fabi et al.
PloS one, 12(5), e0177387-e0177387 (2017-05-06)
Infertility is constantly increasing in Canada, where 16% of Canadian couples are experiencing difficulty conceiving. It is thought that infertility can emanate from the dysregulated communication between the embryo and the maternal endometrium. In order to allow for this window
Use of RNA interference-mediated gene silencing and adenoviral overexpression to elucidate the roles of AKT/protein kinase B isoforms in insulin actions
Katome, T., et al
The Journal of Biological Chemistry, 278, 28312-28323 (2003)
Demetris C Iacovides et al.
Molecular & cellular proteomics : MCP, 12(11), 3210-3220 (2013-08-10)
Breast cancer subtype-specific molecular variations can dramatically affect patient responses to existing therapies. It is thought that differentially phosphorylated protein isoforms might be a useful prognostic biomarker of drug response in the clinic. However, the accurate detection and quantitative analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service