Skip to Content
Merck
All Photos(3)

Documents

05-511

Sigma-Aldrich

Anti-TCF-4 Antibody, clone 6H5-3

clone 6H5-3, Upstate®, from mouse

Synonym(s):

HMG box transcription factor 4, Transcription factor 7-like 2, T-Cell factor 4, T-cell-specific, HMG box, T-Cell-specific transcription factor 4, Transcription factor 7-like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H5-3, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

electrophoretic mobility shift assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TCF7L2(6934)

General description

TCF4, also known as TCF7L2, is expressed widely during development. Gene targeting study indicates that it is required to maintain the crypt stem cells of the small intestine. TCF4 has many different splicing isoforms and they are expressed differentially in tissues and in cancers of different stages. Studies also indicate that variant of the TCF4 gene confers an increased risk of type 2 diabetes.

Specificity

This antibody recognizes TCF-4

Immunogen

Amino acids 31-331 of human TCF-4.

Application

Immunocytochemistry/Immunohistochemistry:
A previous lot of this antibody is reported to show positive immunostaining for TCF-4 in cells fixed with 4% paraformaldehyde/PBS (Amir, A. L., 2003).

Immunoprecipitation:
A previous lot of this antibody was reported to immunoprecipitate TCF-4 from SW620 whole cell lysate as a complex with β-Catenin (Persad, S., 2001).

Supershift Assay:
A previous lot of this antibody was reported to supershift TCF-4:DNA complexes in electrophoretic mobility shift assays (EMSA) (Amir, A. L., 2003).
Use Anti-TCF-4 Antibody, clone 6H5-3 (Mouse Monoclonal Antibody) validated in EMSA, ICC, IHC, IP, WB to detect TCF-4 also known as Immunoglobulin transcription factor 2, SL3-3 enhancer factor 2.

Quality

Routinely evaluated by western blot on RIPA lysates of Cos-1 cells expressing transfected TCF-4.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected TCF-4 in RIPA lysates of Cos-1 cells expressing transfected TCF-4 (Catalog # 21-174).

Target description

Mr 66 kDa

Linkage

Replaces: 04-1080

Physical form

Format: Purified
Purified mouse monoclonal IgG2a in buffer containing 0.07 M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide and 30% glycerol.

Analysis Note

Control
DLD1 (human colorectal adenocarcinoma), Cos-1 cells or HCT116 (human colorectal carcinoma) cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service