Skip to Content
Merck
All Photos(1)

Documents

N-050

Supelco

Nandrolone solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O2
CAS Number:
Molecular Weight:
274.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC12CCC3C(CCC4=CC(=O)CCC34)C1CCC2O

InChI

1S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1

InChI key

NPAGDVCDWIYMMC-IZPLOLCNSA-N

General description

An analytical reference standard applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and urine drug testing to forensic analysis. Nandrolone is an anabolic steroid sold commercially as Deca-Durabolin®. This performance enhancing drug is disallowed in sports competition.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Deca-Durabolin is a registered trademark of Dynamic Sports Nutrition, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M N Kohler et al.
British journal of sports medicine, 36(5), 325-329 (2002-09-28)
The aim of this review is to analyse the studies on nandrolone metabolism to determine if it is possible for an athlete to test positive for nandrolone without having ingested or injected nandrolone.
V Bricout et al.
European journal of applied physiology, 92(1-2), 1-12 (2004-03-26)
Norsteroids are xenobiotics with androgenic and anabolic properties known since as far back as the 1930s. In doping controls, the use of the banned xenobiotic norsteroids is detected in the competitor's urines by the measurement of norandrosterone (19-NA) and noretiocholanolone
[Outcome of osteoporosis in Cushing disease. Presentation of a severe form in adults treated with nandrolone. Very favorable course with a 32-years follow-up].
J C Rénier et al.
Revue du rhumatisme (Ed. francaise : 1993), 61(7-8), 513-520 (1994-01-01)
Adolf E Schindler
The Journal of steroid biochemistry and molecular biology, 142, 48-51 (2013-12-18)
After a worldwide breakdown of hormone therapy [HT] following the publications of the Women's Health Initiative trial and Million Women's Study in 2002-2003, there is now a trend to turn attention again to HT and to explore particular progestogens, which
N K Khankhanian et al.
Military medicine, 157(12), 670-674 (1992-12-01)
We present an unusual pseudotumor that formed in reaction to self-administered intramuscular injections of an anabolic steroid, nandrolone decanoate (Deca-Durabolin) in a young soldier. The histopathologic features which closely mimicked several malignant neoplasms could have led to an incorrect diagnosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service