Skip to Content
Merck
All Photos(1)

Key Documents

A-095

Supelco

Atomoxetine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22ClNO
CAS Number:
Molecular Weight:
291.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CNCC[C@@H](Oc1ccccc1C)c2ccccc2

InChI

1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1

InChI key

LUCXVPAZUDVVBT-UNTBIKODSA-N

Gene Information

human ... SLC6A2(6530)

General description

Atomoxetine is an attention-deficit hyperactivity disorder (ADHD) drug marketed in the US under the trade name Strattera®. This Certified Spiking Solution® is suitable for use in numerous testing applications such as clinical toxicology, forensic analysis, pharmaceutical research, urine drug testing, or calibrator preparation.

Application


  • Metabolic profiling of norepinephrine reuptake inhibitor atomoxetine.: This study investigates the metabolic pathways and profiles of atomoxetine, highlighting its pharmacokinetic properties. The research underscores the significance of Atomoxetine HCl solution in understanding the drug′s metabolism and its implications for ADHD treatment (MacKenzie et al., 2020).


  • Using Cell Cultures for the Investigation of Treatments for Attention Deficit Hyperactivity Disorder.: This systematic review explores the use of cell cultures in studying ADHD treatments, including atomoxetine. It provides insights into the cellular mechanisms and efficacy of Atomoxetine HCl, demonstrating its role in advancing ADHD pharmacological research (Chierrito et al., 2019).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Strattera is a registered trademark of Eli Lilly and Co.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential serious adverse interactions between linezolid and atomoxetine.
Arpit Aggarwal et al.
Journal of child and adolescent psychopharmacology, 22(5), 405-405 (2012-10-23)
Romano Arcieri et al.
Journal of child and adolescent psychopharmacology, 22(6), 423-431 (2013-01-31)
The purpose of this study was to assess the cardiovascular effects of drugs used for attention-deficit/hyperactivity disorder (ADHD) in children and adolescents treated in community care centers in Italy. This study was an open, prospective, observational study of youth with
M Reza Mirbolooki et al.
Synapse (New York, N.Y.), 67(2), 79-93 (2012-10-20)
Brown adipose tissue (BAT) plays a significant role in metabolism. In this study, we report the use of atomoxetine (a clinically applicable norepinephrine reuptake inhibitor) for (18)F-FDG PET imaging of BAT and its effects on heat production and blood glucose
F Livio
Revue medicale suisse, 9(368), 72-75 (2013-02-02)
Main pharmacovigilance updates in 2012 are reviewed here. Dabigatran: elderly patients with renal failure are at higher risk of bleeding. Dual renin-angiotensin-aldosterone system blockade comprising aliskiren is harmful. Incretins: low risk of acute pancreatitis. Interaction between fusidic acid and statins:
Hedi Schelleman et al.
PloS one, 8(1), e52991-e52991 (2013-02-06)
To compare the incidence rates of serious cardiovascular events in adult initiators of amphetamines or atomoxetine to rates in non-users. This was a retrospective cohort study of new amphetamines (n=38,586) or atomoxetine (n=20,995) users. Each medication user was matched to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service