Skip to Content
Merck
All Photos(1)

Key Documents

878110P

Avanti

C16-02:0 PC

Avanti Research - A Croda Brand 878110P, powder

Synonym(s):

1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54NO7P
CAS Number:
Molecular Weight:
523.68
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (878110P-25mg)
pkg of 1 × 5 mg (878110P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878110P

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C26H54NO7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-31-23-26(34-25(2)28)24-33-35(29,30)32-22-20-27(3,4)5/h26H,6-24H2,1-5H3/t26-/m1/s1

InChI key

HVAUUPRFYPCOCA-AREMUKBSSA-N

General description

C16-02:0 PC, also referred as 1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine (C16 PAF) (acetyl PAF), is a platelet-activating factor (PAF). Acetyl PAF is a natural PAF with ether-linked hexadecyl chain at the sn-1 position and an acetyl group at the sn-2 position.

Application

C16-02:0 PC or 1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine may be used as a substrate in enzymatic lipoprotein-associated phospholipase A2 (Lp-PLA2) activity assay. It may also be used to study its physicochemical properties and its interaction with peptides derived from the C-terminal domains of human apolipoprotein E.

Biochem/physiol Actions

C16-02:0 PC, a platelet-activating factor and a lipid mediator is involved in various inflammatory diseases. It facilitates initiation and progression of atherosclerosis. PAF acts as a potential biomarker for coronary artery diseases. It exhibits its activity by binding to a specific PAF receptor, which belongs to the seven transmembrane-spanning G-protein-linked receptors family. PAF on hydrolysis by platelet-activating factor-acetyl hydrolase produces LysoPAF (1-O-alkyl-2-hydroxy-sn-glycero-3-phosphocholine).

Packaging

5 mL Clear Glass Sealed Ampule (878110P-25mg)
5 mL Clear Glass Sealed Ampule (878110P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Sato et al.
European journal of pharmacology, 745, 46-51 (2014-12-03)
Platelet-activating factor (PAF) is a potent lipid mediator that is implicated in numerous inflammatory diseases. C-reactive protein (CRP) is an acute-phase plasma protein that increases rapidly and dramatically in response to inflammation. In this study, we investigated the effect of
Sunil A Nankar et al.
Biochimie, 95(6), 1196-1207 (2013-02-05)
Apolipoprotein-derived peptides are promising candidates for the treatment of various inflammatory conditions and the main mechanism proposed for the protective action of these peptides includes binding to pro-inflammatory lipid mediators with high affinity and facilitating their sequestration/metabolism/clearance in the body.
Saki Yamaura et al.
Clinica chimica acta; international journal of clinical chemistry, 481, 184-188 (2018-03-20)
Measurement of lipoprotein-associated phospholipase A2 (Lp-PLA2) can be used as an adjunct to traditional cardiovascular risk factors for identifying individuals at higher risk of cardiovascular events. This can be performed by quantification of the protein concentration using an ELISA platform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service