Skip to Content
Merck
All Photos(2)

Documents

840445P

Avanti

14:0 PG

Avanti Research - A Croda Brand

Synonym(s):

1,2-ditetradecanoyl-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); DMPG; PG(14:0/14:0); 110648, DMPG sodium salt, Dimyristoylphosphatidylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H66O10PNa
CAS Number:
Molecular Weight:
688.85
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840445P-1g)
pkg of 1 × 200 mg (840445P-200mg)
pkg of 1 × 25 mg (840445P-25mg)
pkg of 1 × 500 mg (840445P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C34H67O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-33(37)41-29-32(30-43-45(39,40)42-28-31(36)27-35)44-34(38)26-24-22-20-18-16-14-12-10-8-6-4-2;/h31-32,35-36H,3-30H2,1-2H3,(H,39,40);/q;+1/p-1/t31?,32-;/m1./s1

InChI key

QLNOOKSBAYIHQI-SKZICHJRSA-M

General description

Phosphoglyceride (PG) are also referred as glycerophospholipid. It is an important component of membrane bilayers. It has three-carbon backbone of glycerol, two long-chain fatty acids, esterified to hydroxyl groups on carbons 1 and 2 of the glycerol and phosphoric acid esterified to the C3 hydroxyl group of glycerol. It is found at high levels in the membranes of all cells and at low levels in fat stores.

Application

14:0 PG is suitable for use:
  • in the preparation of lipid bicelles for nuclear magnetic resonance (NMR) experiment
  • as phospholipid (PL) standard to quantify phosphatidylglycerol (PG) by chromatography
  • for the preparation of DMPG (l-dimyristoylphosphatidylglycerol) phospholipid vesicles

Biochem/physiol Actions

Phosphoglycerides (PG)/glycerophospholipids play a key role in cell signaling systems. It also serves as an anchor for proteins in cell membranes.

Packaging

20 mL Clear Glass Screw Cap Vial (840445P-1g)
20 mL Clear Glass Screw Cap Vial (840445P-500mg)
5 mL Amber Glass Screw Cap Vial (840445P-200mg)
5 mL Amber Glass Screw Cap Vial (840445P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction and profiling of plant polar glycerol lipids
Liu Y and Wang X
Bio-protocol, 10 (2016)
Lipids
Medical Biochemistry, 10, 99- 119 (2017)
Lipid binding by disordered proteins
Perez Y, et al.
Protocol Exchange, 10 (2013)
Lipid binding by disordered proteins
Perez Y, et al.
Protocol Exchange, 10 (2013)
In vitro Membrane Interaction and Liposome Fusion Assays Using Recombinant Hepatitis C Virus Envelope Protein E2
Yelamos B, et al.
Biochimica et Biophysica Acta - Biomembranes, 10 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service