Key Documents
Z645192
IKA® A 11.3 hard metal beater
A 11.3 beater, coated with chromium carbide, 1/cs
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
material
stainless steel
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 2983000
suitability
suitable for (pulverizing substances with a Mohs hardness up to 9)
General description
Stainless stell rackets for use with the IKA A11 Basic Analytical Mill, for breaking hard substances up to Mohs hardness 9,
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)