Z169609
Spectroline® E-Series UV lamp
output4 W, AC/DC input 230 V AC
Synonym(s):
TM=["Spectroline"] E-Series Ultraviolet Light
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
thin layer chromatography (TLC): suitable
W × L × H
3 1/4 in. × 8 in. × 2 1/2 in.
output
(This product is supplied with an EU plug)
4 W
Looking for similar products? Visit Product Comparison Guide
General description
May be used alone (hand-held) or in viewing cabinets.
Lamps have one longwave (365 nm) and one shortwave (254 nm) tube, 115V
Lamps have one longwave (365 nm) and one shortwave (254 nm) tube, 120V
Lamps have one longwave (365 nm) and one shortwave (254 nm) tube, 230 V
Lamps have one longwave (365 nm) and one shortwave (254 nm) tube, 115V
Lamps have one longwave (365 nm) and one shortwave (254 nm) tube, 120V
Lamps have one longwave (365 nm) and one shortwave (254 nm) tube, 230 V
Legal Information
Spectroline is a registered trademark of Spectronics, Corp.
replacement part
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service