Skip to Content
Merck
All Photos(2)

Key Documents

G900

Sigma-Aldrich

Girard′s reagent T

98%

Synonym(s):

(Carboxymethyl)trimethylammonium chloride hydrazide, (Hydrazinocarbonylmethyl)trimethylammonium chloride, Acethydrazide trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3N+CH2CONHNH2]Cl-
CAS Number:
Molecular Weight:
167.64
Beilstein:
3727163
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Assay

98%

mp

188-192 °C (dec.) (lit.)

SMILES string

[Cl-].C[N+](C)(C)CC(=O)NN

InChI

1S/C5H13N3O.ClH/c1-8(2,3)4-5(9)7-6;/h4,6H2,1-3H3;1H

InChI key

YSULOORXQBDPCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Girard′s reagent T, also known as (Carboxymethyl)trimethylammonium chloride hydrazide, is a cationic surfactant that can be used as a derivatizing agent for ketones and aldehydes to form hydrazones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David W Johnson
Rapid communications in mass spectrometry : RCM, 21(18), 2926-2932 (2007-08-07)
4-Hydrazino-N,N,N-trimethyl-4-oxobutanaminium iodide (HTMOB) is a modified Girard derivatizing reagent synthesized to improve the sensitivity of analysis of aldehydes and ketones with electrospray ionization tandem mass spectrometry. Compared with Girard T reagent the measured signal intensity increase is between 3.3 times
Usama El-Ayaan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(2), 211-219 (2007-02-24)
A new series of first-row transition metal complexes with 1-acetylpyridinium chloride-4-benzoyl thiosemicarbazide (H2GPBzIT) have been prepared and characterized by elemental analysis, spectroscopic and magnetic measurements. The proton-ligand ionization constants were determined potentiometrically using Irving-Rossotti technique. The stability constants of complexes
Kyoung-Soon Jang et al.
Analytical biochemistry, 386(2), 228-236 (2009-01-13)
Quality control and assurance of glycan profiles of a recombinant glycoprotein from lot to lot is a critical issue in the pharmaceutical industry. To develop an easy and simple quantitative and qualitative glycan profile method based on matrix-assisted laser desorption/ionization-time-of-flight
M M Mostafa
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(2), 480-486 (2006-08-30)
Three new derivatives of thiosemicarbazides, derived from the reactions of ethyl-, allyl- and phenyl isothiocyanates with Girard's T [(carboxymethyl) trimethylammonium chloride hydrazide] and P [1-(carboxymethyl) pyridinium chloride hydrazide], have been isolated and characterized by chemical analyses, conductivities, spectral (IR, 1H
S Görög et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 1115-1124 (1996-06-01)
4-Ene-3-ketosteroids and 17-ketosteroids were quantitatively transformed into the corresponding hydrazones using Girard P and T reagents, respectively. The positively charged derivatives were separated by capillary electrophoresis. The spectrophotometric characteristics of the derivatives permitted their sensitive detection in the 230-280 nm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service