Skip to Content
Merck
All Photos(3)

Documents

B33803

Sigma-Aldrich

Bicyclo[2.2.1]hepta-2,5-diene

98%

Synonym(s):

2,5-Norbornadiene, NBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8
CAS Number:
Molecular Weight:
92.14
Beilstein:
506224
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

contains

0.05-0.25% BHT as inhibitor

refractive index

n20/D 1.470 (lit.)

bp

89 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

SMILES string

C1[C@H]2C=C[C@@H]1C=C2

InChI

1S/C7H8/c1-2-7-4-3-6(1)5-7/h1-4,6-7H,5H2/t6-,7+

InChI key

SJYNFBVQFBRSIB-KNVOCYPGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate in prostaglandin synthesis.

Features and Benefits

Intermediate in prostaglandin synthesis.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-5.8 °F - closed cup

Flash Point(C)

-21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Victoria Jiménez et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(29), 8115-8128 (2011-06-03)
The treatment of [{Rh(μ-SH){P(OPh)(3)}(2)}(2)] with [{M(μ-Cl)(diolef)}(2)] (diolef=diolefin) in the presence of NEt(3) affords the hydrido-sulfido clusters [Rh(3)(μ-H)(μ(3)-S)(2)(diolef){P(OPh)(3)}(4)] (diolef=1,5-cyclooctadiene (cod) for 1, 2,5-norbornadiene (nbd) for 2, and tetrafluorobenzo[5,6]bicyclo[2.2.2]octa-2,5,7-triene (tfb) for 3) and [Rh(2)Ir(μ-H)(μ(3)-S)(2)(cod){P(OPh)(3)}(4)] (4). Cluster 1 can be also obtained by
Paulo M Nunes et al.
The journal of physical chemistry. A, 113(23), 6524-6530 (2009-05-20)
The energetics of tert-butoxyl radical addition reaction to norbornadiene was investigated by time-resolved photoacoustic calorimetry (TR-PAC). The result, together with the C-O bond dissociation enthalpy (BDE) in the addition product, allowed us to calculate the pi-bond dissociation enthalpy in norbornadiene.
Tanner J Kettles et al.
The Journal of organic chemistry, 76(16), 6951-6957 (2011-07-08)
Ruthenium-catalyzed homo Diels-Alder [2 + 2 + 2] cycloadditions between alkynyl phosphonates and bicyclo[2.2.1]hepta-2,5-diene were studied. The observed reactivity was found to be dependent on the presence of the phosphonate moiety. The Ru-catalyzed cycloaddition was compatible with a variety of
Yusuke Kamiyoshihara et al.
Plant physiology, 160(1), 488-497 (2012-07-17)
Perception of the plant hormone ethylene is essential to initiate and advance ripening of climacteric fruits. Since ethylene receptors negatively regulate signaling, the suppression is canceled upon ethylene binding, permitting responses including fruit ripening. Although receptors have autophosphorylation activity, the
Ehud Katz et al.
Journal of experimental botany, 56(415), 1359-1367 (2005-03-16)
Although leaves and other vegetative tissues are generally considered as non-climacteric, citrus leaves show a climacteric system II behaviour after detachment. Upon harvest, young, fully expanded 'Valencia' orange (Citrus sinensis) leaves ( approximately 60-d-old) exhibited two phases of ethylene production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service