Skip to Content
Merck
All Photos(2)

Key Documents

935913

Sigma-Aldrich

Lithium tetrafluoroborate

greener alternative

Technipur®, ultra dry, powder, 99.99% trace metals basis

Synonym(s):

Lithium borofluoride, Lithium fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBF4
CAS Number:
Molecular Weight:
93.75
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

Quality Level

Assay

99.99% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

293-300 °C (dec.) (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].F[B-](F)(F)F

InChI

1S/BF4.Li/c2-1(3,4)5;/q-1;+1

InChI key

UFXJWFBILHTTET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product serves as an electrolyte with excellent ionic conductivity and is commonly used in the production of lithium-ion batteries, thus it has been enhanced for energy efficiency. Click here for more information.

Application

Lithium tetrafluoroborate (LiBF4 ) is used as a solid composite electrolyete for lithium ion batteries. High ionic conductivity, greater thermal stability makes it a good candidate for battery electrolyte.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrolyte solvents for high voltage lithium ion batteries: ion correlation and specific anion effects in adiponitrile
Kirshnamoorthy, A.N.
Physical Chemistry Chemical Physics, 20 (2018)
Electrical, dielectric and electrochemical characterization of novel poly(acrylic acid)-based polymer electrolytes complexed with lithium tetrafluoroborate
Ngai, Koh Sing
Chemical Physics Letters, 692 (2018)
Investigation of the synergetic effects of LiBF4 and LiODFB as wide-temperature electrolyte salts in lithium-ion batteries
Zhang, L.
Ionics, 24 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service