Skip to Content
Merck
All Photos(1)

Key Documents

933139

Sigma-Aldrich

SEM Sample Holder

3-Pin Configuration Holder

Synonym(s):

3-pin SEM mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

Looking for similar products? Visit Product Comparison Guide

General description

EMS Techware

Application

Our specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins. Be sure to order the Adapter Pin that fits your instrument. For this reason, we offer three different types of pin adapters, which are threaded and ready to screw on to the base of the holders.
All adapters have a threaded portion 5 mm in length.
The 3-Pin configuration holder is designed to save time. It accommodates 3 of ½" (12.5 mm) diameter surface, ⅛" (3.1 mm) diameter pin.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service