Skip to Content
Merck
All Photos(1)

Key Documents

900354

Sigma-Aldrich

Ultroxa®: Poly(2-methyl-2-oxazoline) azide terminated

average Mn 10,000, PDI ≤1.2

Synonym(s):

PMeOx-Azide, PMeOx-N3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C4H7NO)nN3
UNSPSC Code:
12352125
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn 10,000

PDI

≤1.2

storage temp.

2-8°C

Legal Information

Ultroxa is a registered trademark of University of Ghent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service