Skip to Content
Merck
All Photos(1)

Key Documents

860255

Sigma-Aldrich

6-Fluorotryptamine hydrochloride

99%

Synonym(s):

3-(2-Aminoethyl)-6-fluoroindole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11FN2 · HCl
CAS Number:
Molecular Weight:
214.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

233-235 °C (lit.)

SMILES string

Cl.NCCc1c[nH]c2cc(F)ccc12

InChI

1S/C10H11FN2.ClH/c11-8-1-2-9-7(3-4-12)6-13-10(9)5-8;/h1-2,5-6,13H,3-4,12H2;1H

InChI key

ODVDIBHWTMTHOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Indole alkaloid drug candidates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Demosthenes Fokas et al.
Molecular diversity, 9(1-3), 81-89 (2005-03-26)
A series of diverse indole-based chemotypes were synthesized from beta-tetrahydrocarboline (beta-THC) scaffolds prepared from commercially and readily available tryptamines and alpha-ketoesters. Diversity can be generated within these chemotypes through the following strategies: (a) appendage of substituents to the beta-THC scaffold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service