Skip to Content
Merck
All Photos(1)

Key Documents

772577

Sigma-Aldrich

Polystyrene, DDMAT terminated

average Mn 5,000, PDI <1.1

Synonym(s):

Poly(styrene) RAFT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7O2(C8H8)nC13H25S3
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

average Mn 5,000

transition temp

Tm 361-366 °C

PDI

<1.1

Looking for similar products? Visit Product Comparison Guide

Application

Poly(styrene) is a hydrophobic polymer used in many applications. The end group functionality contains a carboxylic acid (COOH) as well as the 2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid RAFT agent moiety. This will allow for additional polymerization to generate a diblock copolymer. This RAFT agent is especially suited for the polymerization of styrene; acrylate and acrylamide monomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service