Skip to Content
Merck
All Photos(2)

Key Documents

764175

Sigma-Aldrich

P(Cy3) Pd G3

97%

Synonym(s):

Palladium G3-Tricyclohexylphosphine, [(Tricyclohexylphosphine)-2-(2′-aminobiphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46NO3PPdS
CAS Number:
Molecular Weight:
650.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

219-224 °C (decomposition)

functional group

phosphine

SMILES string

CS(=O)(=O)O[Pd]c1ccccc1-c2ccccc2N.C3CCC(CC3)P(C4CCCCC4)C5CCCCC5

InChI

1S/C18H33P.C12H10N.CH4O3S.Pd/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h16-18H,1-15H2;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

HZVUEHVNPWCEIW-UHFFFAOYSA-M

General description

P(Cy3) Pd G3 is a 3rd generation phosphine functionalized palladium precatalyst used in organic synthesize, that is often used in cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of gem-Difluorinated 1, 3-Dienes via Synergistic Cu/Pd-Catalyzed Borodifluorovinylation of Alkynes
Xu WY, et al.
Organic Letters, 24(32), 5884-5889 (2022)
Catalyst-Controlled 1, 2-and 1, 1-Arylboration of ?-Alkyl Alkenyl Arenes
Bergmann AM, et al.
Angewandte Chemie (International Edition in English), 131(6), 1733-1737 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service