Skip to Content
Merck
All Photos(2)

Documents

760773

Sigma-Aldrich

Dibenzocyclooctyne-PEG4-Fluor 545

for Copper-free Click Chemistry

Synonym(s):

Polyethylene glycol, ADIBO-PEG4-TAMRA, DBCO-PEG4-Fluor 545, DBCO-PEG4-TAMRA, DBCO-PEG4-tetramethylrhodamine, TAMRA DBCO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H57N5O10
Molecular Weight:
936.06
UNSPSC Code:
51171641
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

Application

This azadibenzocyclooctyne-tetramethyl rhodamine (TAMRA) derivative is a versatile fluorescent reagent for labeling azide containing molecules or compounds. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne click chemistry reactions.This dibenzocyclooctyne will react with azide-functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. Subsequent labeled molecules can be visualized by fluorescence spectroscopy.

Spectral Properties: Abs/Em = 545/567nm

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning, X., et al.
Angewandte Chemie (International Edition in English), 47, 2253-2255 null
Rubino F. A., et al.
Journal of Visualized Experiments, 66, 4246-4246 null
Accelerated Strain-Promoted and Oxidation-Controlled Cyclooctyne-Quinone Cycloaddition for Cell Labeling.
George A, et al.
ChemistrySelect, 2(24), 7117-7122 null
Single-step synthesis of small, azide-functionalized gold nanoparticles: Versatile, water-dispersible reagents for click chemistry.
Elliott EW, et al.
Langmuir (2017)
John T Ngo et al.
ACS chemical biology, 7(8), 1326-1330 (2012-06-14)
Transcriptional activity from a specified promoter can provide a useful marker for the physiological state of a cell. Here we introduce a method for selective tagging of proteins made in cells in which specified promoters are active. Tagged proteins can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service