Skip to Content
Merck
All Photos(1)

Documents

755737

Sigma-Aldrich

Tungsten hexacarbonyl

packaged for use in deposition systems

Synonym(s):

Tungsten hexacarbonyl, Hexacarbonyltungsten (0), Tungsten carbonyl, Tungstenhexacarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
W(CO)6
CAS Number:
Molecular Weight:
351.90
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

12.1 (vs air)

vapor pressure

1.2 mmHg ( 67 °C)

Assay

99.9% (trace metals analysis)

form

solid

purified by

sublimation

reaction suitability

core: tungsten

mp

150 °C (lit.)

density

2.65 g/mL at 25 °C (lit.)

SMILES string

[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/6CO.W/c6*1-2;

InChI key

FQNHWXHRAUXLFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tumgstenhexacarbonyl is a volatile, air stable precursor of Tungsten(0), widely used in ALD and CVD techniques.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mulders, J. J. L.; Belova, L. M.; Riazanova, A.
Nanotechnology, 22, 055302/1-055302/1 (2011)
Maruyama, T., Arai, S.
Journal of the Electrochemical Society, 4, 1021-1021 (1994)
S Martin et al.
The Journal of chemical physics, 134(3), 034304-034304 (2011-01-26)
We have studied the excitation and dissociation processes of the molecule W(CO)(6) in collisions with low kinetic energy (3 keV) protons, monocharged fluorine, and chlorine ions using double charge transfer spectroscopy. By analyzing the kinetic energy loss of the projectile
F Porrati et al.
Nanotechnology, 20(19), 195301-195301 (2009-05-08)
W-based granular metals have been prepared by electron-beam-induced deposition from the tungsten hexacarbonyl W(CO)(6) precursor. In situ electrical conductivity measurements have been performed to monitor the growth process and to investigate the behavior of the deposit under electron beam post-irradiation
Lei Li et al.
Journal of the American Society for Mass Spectrometry, 15(1), 87-102 (2003-12-31)
The internal energy distributions, P(epsilon), of a millisecond pulsed radio frequency glow discharge plasma were investigated using tungsten hexcarbonyl W(CO)(6) as a "thermometer molecule". Vapor of the probe molecule, W(CO)(6), was introduced into the plasma and subjected to various ionization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service