Skip to Content
Merck
All Photos(1)

Key Documents

75056

Sigma-Aldrich

(+)-Valencene

technical, ≥70%

Synonym(s):

(3R,4aS,5R)-4a,5-Dimethyl-3-isopropenyl-1,2,3,4,4a,5,6,7-octahydronaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
3539153
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥70%

form

liquid

optical activity

[α]20/D +100±25°, neat

refractive index

n20/D 1.504 (lit.)

bp

274 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1CCC=C2CC[C@H](C[C@@]12C)C(C)=C

InChI

1S/C15H24/c1-11(2)13-8-9-14-7-5-6-12(3)15(14,4)10-13/h7,12-13H,1,5-6,8-10H2,2-4H3/t12-,13-,15+/m1/s1

InChI key

QEBNYNLSCGVZOH-NFAWXSAZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Valencene is a sesquiterpene, which contains decalin framework and two methyl-bearing stereocenters. It is an aroma component of citrus fruit.

Application

(+)-Valencene can be used as a precursor to prepare:
  • (+)-Nootkatone (a sesquiterpene) by dark singlet oxygenation.
  • Benzoyloxyvalencene by reacting with tert-butyl peroxy benzoate via Kharasch−Sosnovsky allylic oxidation method.
  • (+)-Lineariifolianone, a natural product.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-Pot Synthesis of (+)-Nootkatone via Dark Singlet Oxygenation of Valencene: The Triple Role of the Amphiphilic Molybdate Catalyst.
Hong B, et al.
Catalysts, 6(12), 184-184 (2016)
Optimization by Response Surface Methodology (RSM) of the Kharasch-Sosnovsky Oxidation of Valencene
Garcia-Cabeza AL, et al.
Organic Process Research & Development, 19(11), 1662-1666 (2015)
CitAP2. 10 activation of the terpene synthase CsTPS1 is associated with the synthesis of (+)-valencene in 'Newhall'orange.
Shen SL, et al.
Journal of Experimental Botany, 67(14), 4105-4115 (2016)
One-pot synthesis of (+)-nootkatone via dark singlet oxygenation of valencene: The triple role of the amphiphilic molybdate catalyst
Hong B, et al.
Catalysts, 6(12), 184-184 (2016)
Synthesis of (+)-Lineariifolianone and Related Cyclopropenone-Containing Sesquiterpenoids
Reber KP, et al.
The Journal of Organic Chemistry, 84(9), 5524-5534 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service