Skip to Content
Merck
All Photos(1)

Key Documents

741884

Sigma-Aldrich

Poly(2-ethyl-2-oxazoline)

average Mn 20,000, PDI <1.4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 20,000

mp

>300 °C

PDI

<1.4

Looking for similar products? Visit Product Comparison Guide

Application

This Poly(2-ethyl-2-oxazoline) polymer is amorphous and water soluble with good temperature stability. Jordan and coworkers showed biocompatibility, no accumulation in tissue, and rapid clearance from the bloodstream1. End-group modified poly(2-ethyl-2-oxazoline)s have been conjugated to peptides2, and were shown as versatile alternatives to poly(ethylene glycol) (PEG) for both protein and small drug conjugation3.

Potential substitute for poly(vinyl alcohol) and poly(vinyl pyrrolidone). Sometimes it is used as an adhesion promoter in coatings. This is also sometimes used in heat sealing and remoistenable hot-melt adhesives. N-propionyl substituted linear polyethylenimine

Features and Benefits

Nonionic, water-soluble thermoplastic. Better heat stability than poly(vinyl alcohol). Good melt flow, shear stability and Newtonian characteristics. Water is a room temperature Theta solvent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Luxenhofer, M. Lopez-Garcia, A. Frank, H. Kessler, R. Jordan
Polym. Mater. Sci. Eng., 95, 283-284 (2006)
A. Mero, G. Pasut, L. D. Via, M.W. M. Fijten, et al.
J. Controlled Release, 125, 87-95 (2008)
F. C. Gaertner, R. Luxenhofer, B. Blechert, R. Jordan, M. Essler
J. Controlled Release, 119, 291-300 (2007)
J H Jeong et al.
Journal of controlled release : official journal of the Controlled Release Society, 73(2-3), 391-399 (2001-08-23)
A series of linear poly(ethylenimine) (L-PEI) containing varying amounts of cationic charge density in its backbone was produced by controlled hydrolysis of poly(2-ethyl-2-oxazoline) (PEtOz) for using as a nonviral DNA transfection agent. The effects of cationic charge density and molecular
Issa Katime et al.
Applied spectroscopy, 57(7), 829-834 (2003-12-09)
This paper reports an FT-IR study of blends of poly(mono-n-alkyl itaconates) with poly(N,N-dimethylacrylamide) (PDMA) and poly(ethyloxazoline) (PEOX). Strong hydrogen bonding has been found, and both polybases have shown similar acceptor strengths. Derivative techniques show asymmetric profiles for the free carbonyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service