Skip to Content
Merck
All Photos(3)

Key Documents

734578

Sigma-Aldrich

Ruthenium iodide

anhydrous, ≥99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I3Ru
CAS Number:
Molecular Weight:
481.78
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

Assay

≥99% trace metals basis

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

590 °C (dec.)

SMILES string

I[Ru](I)I

InChI

1S/3HI.Ru/h3*1H;/q;;;+3/p-3

InChI key

LJZVDOUZSMHXJH-UHFFFAOYSA-K

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service