719854
Resomer® L 206 S, Poly(L-lactide), ester terminated
Synonym(s):
PLL
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
granular
degradation timeframe
>3 years
viscosity
0.8-1.2 dL/g, 0.1 % in chloroform(25 °C, Ubbelohde) (size 0c glass capillary viscometer)
transition temp
Tm 180-185 °C
Tg 60-65 °C
storage temp.
2-8°C
General description
Resomer® L 206 S, Poly(L-lactide) (PLLA) is a high-performance biodegradable polymer with an ester-terminated end group. Its properties include good mechanical stability, hydrophobicity, and excellent composting characteristics. It is a non-toxic, biocompatible, and bioresorbable polymer widely used in biomedical applications.
Application
Resomer® L 206 S, Poly(L-lactide) (PLLA) is an optically active and semi-crystalline material that can be used in the fabrication of biomedical devices and research tissue engineering solutions, such as orthopedics, soft tissue fixation devices, sports medicine, plates, screws, nails, and stents. This bioresorbable material exhibits a resorption period greater than 3 years whereas the inherent viscosity is 0.8–1.2. The advancement in biomedical applications such as osteoconductivity of these composite materials has improved by the introduction of calcium-based inorganic additives.
Legal Information
Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis and properties of Poly (L-lactide)-Poly (ε-caprolactone) multiblock copolymers by the self-polycondensation of diblock macromonomers
Polymer Journal, 47(10), 657-665 (2015)
Blends of aliphatic polyesters: V non-enzymatic and enzymatic hydrolysis of blends from hydrophobic poly (l-lactide) and hydrophilic poly (vinyl alcohol)
Polymer Degradation and Stability, 71(3), 403-413 (2001)
Bioresorbable Materials for Orthopedic Applications (Lactide and Glycolide Based)
Orthopedic Biomaterials (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service