Skip to Content
Merck
All Photos(1)

Documents

719234

Sigma-Aldrich

1-Adamantanethiol

99% (GC)

Synonym(s):

ADT, 1-AD, 1-AdSH, 1-AdmSH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16S
CAS Number:
Molecular Weight:
168.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (GC)

form

solid

mp

103-105 °C

SMILES string

SC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C10H16S/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9,11H,1-6H2/t7-,8+,9-,10-

InChI key

ADJJLNODXLXTIH-CHIWXEEVSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xi Kang et al.
Chemical science, 9(11), 3062-3068 (2018-05-08)
The strategy of aggregation-induced emission (AIE) has been widely used to enhance the photo-luminescence (PL) in the nanocluster (NC) research field. Most of the previous reports on aggregation-induced enhancement of fluorescence in NCs are induced by the restriction of intramolecular
Matthew L Gethers et al.
ACS nano, 9(11), 10909-10915 (2015-10-02)
We demonstrate the use of "holey" graphene as a mask against molecular adsorption. Prepared porous graphene is transferred onto a Au{111} substrate, annealed, and then exposed to dilute solutions of 1-adamantanethiol. In the pores of the graphene lattice, we find
Dennis B Pacardo et al.
Nanoscale, 7(28), 12096-12103 (2015-07-01)
A multifunctional gold nanorod (AuNR) complex is described with potential utility for theranostic anticancer treatment. The AuNR was functionalized with cyclodextrin for encapsulation of doxorubicin, with folic acid for targeting, and with a photo-responsive dextran-azo compound for intracellular controlled drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service