Skip to Content
Merck
All Photos(1)

Key Documents

694290

Sigma-Aldrich

Tris(2-chloroethyl)phosphate-d12

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCD2CD2O)3P(O)
Molecular Weight:
297.56
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

bp

192 °C/10 mmHg (lit.)

density

1.448 g/mL at 25 °C

mass shift

M+12

SMILES string

[2H]C([2H])(Cl)C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])Cl)OC([2H])([2H])C([2H])([2H])Cl

InChI

1S/C6H12Cl3O4P/c7-1-4-11-14(10,12-5-2-8)13-6-3-9/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2

InChI key

HQUQLFOMPYWACS-LBTWDOQPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

449.6 °F - closed cup

Flash Point(C)

232 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang-Ying Liu et al.
Journal of chromatography. A, 1406, 251-257 (2015-07-01)
A method for the digestion, extraction, fractionation, and analysis of three classes of flame retardants, including 36 polybrominated diphenyl ethers (PBDEs), 9 halogenated alternative flame retardants (AFRs), and 12 organophosphate esters (OPEs) in human hair and nail samples was developed.
Angela A Peverly et al.
Environmental science & technology, 48(19), 11154-11160 (2014-09-12)
The Michigan Chemical (also known as Velsicol Chemical) plant located in St. Louis, Michigan operated from 1936-1978. During this time, the plant manufactured polybrominated biphenyls (PBBs), hexabromobenzene (HBB), 1,1,1-trichloro-2,2-bis(4-chlorophenyl) ethane (DDT), and tris(2,3-dibromopropyl) phosphate (TDBPP), among other products. Due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service