Skip to Content
Merck
All Photos(2)

Key Documents

685976

Sigma-Aldrich

Mischmetal-nickel alloy

greener alternative

hydrogen-storage grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Ce, La, Nd, Pr)Ni5
CAS Number:
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

hydrogen-storage grade

description

Hydrogen Storage Capacity: 1.5-1.6 wt.% @25°C

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Mischmetal-nickel alloy is a hydrogen storage alloy, which consists of rare earth metals and transition metals. It can be used as an anode material for the formation of nickel-metal hydride batteries. These batteries can further be utilized in hybrid vehicles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Pyr. Sol. 1 - Repr. 1B - Resp. Sens. 1 - Self-heat. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - Water-react 1

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recovery of valuable metals from a spent nickel-metal hydride battery: Selective chlorination roasting of an anodic active material with CCl4 gas
Kuzuya T, et al.
Separation and Purification Technology, 118, 823-827 (2013)
Structural, hydrogen storage and thermodynamic properties of some mischmetal-nickel alloys with partial substitutions for nickel
Kumar EA, et al.
Journal of alloys and compounds, 476(1-2), 92-97 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service